Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ADCY10 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | ADCY10 |
---|---|
Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 |
Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
ADCY10 Polyclonal specifically detects ADCY10 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
ADCY10 | |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Human | |
adenylate cyclase 10 (soluble), Adenylate cyclase homolog, adenylate cyclase type 10, AH-related protein, ATP pyrophosphate-lyase, EC 4.6.1.1, Germ cell soluble adenylyl cyclase, HCA2RP1-313L4.2, hsAC, sAC, SAC3'-5'-cyclic AMP synthetase, SACI, Sacy, Testicular soluble adenylyl cyclase, testicular soluble adenylyl cyclase (SAC) | |
ADCY10 | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
Polyclonal | |
Rabbit | |
Lipid and Metabolism | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
55811 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:NLENLVAQNTTGPVFCPRLYHLMAYVCILMGDGQKCGLFLNTALRLSETQGNILEKCWLNMNKESWYSTSELKEDQWLQTILSLPSW | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title