Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ADCY2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA531284
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA531284 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA531284 Supplier Invitrogen™ Supplier No. PA531284
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: NT2D1, IMR32, U87-MG, MCF-14. Predicted reactivity: Mouse (97%), Rat (97%), Rhesus Monkey (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

ADCY2 is a member of the family of adenylate cyclases, which are membrane-associated enzymes that catalyze the formation of the secondary messenger cyclic adenosine monophosphate(cAMP). This enzyme is insensitive to Ca(2+)/calmodulin, and is stimulated by the G protein beta and gamma subunit complex.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ADCY2
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Gene ADCY2
Gene Accession No. Q08462
Gene Alias 3',5'-cyclic AMP synthetase; AC2; Adcy2; adenylate cyclase 2; adenylate cyclase 2 (brain); adenylate cyclase II; adenylate cyclase type 2; Adenylate cyclase type II; adenylyl cyclase 2; ATP pyrophosphate-lyase 2; HBAC2; KIAA1060; mKIAA1060; type II adenylate cyclase; type II adenylyl cyclase
Gene Symbols ADCY2
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the C-terminus region of human Adenylate cyclase 2. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 108
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.