Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ADCY2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597247
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597247 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597247 Supplier Invitrogen™ Supplier No. PA597247
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VTLEHLNGAY KVEEGDGDIR DPYLKQHLVK TYFVINPKGE RRSPQHLFRP RHTLDGAKMR ASVRMTRYLE SWGAAKPFAH LHHRDSMTTE NGKISTTDVP MGQHNFQNRT LRTKSQKKRF EEELNERMIQ AIDGINAQKQ WLKSEDIQRI SLLFYNKVLE KEYRATALPA F; Positive Samples: SH-SY5Y, Rat eye; Cellular Location: Cell membrane, Cytoplasm, Membrane, Multi-pass membrane protein.

ADCY2 is a member of the family of adenylate cyclases, which are membrane-associated enzymes that catalyze the formation of the secondary messenger cyclic adenosine monophosphate(cAMP). This enzyme is insensitive to Ca(2+)/calmodulin, and is stimulated by the G protein beta and gamma subunit complex.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ADCY2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.46 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ADCY2
Gene Accession No. P26769, Q08462, Q80TL1
Gene Alias 3',5'-cyclic AMP synthetase; AC2; Adcy2; adenylate cyclase 2; adenylate cyclase 2 (brain); adenylate cyclase II; adenylate cyclase type 2; Adenylate cyclase type II; adenylyl cyclase 2; ATP pyrophosphate-lyase 2; HBAC2; KIAA1060; mKIAA1060; type II adenylate cyclase; type II adenylyl cyclase
Gene Symbols ADCY2
Host Species Rabbit
Immunogen Recombinant protein (or fragment)
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 108, 210044, 81636
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.