Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ADCY4 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP23792025UL
Description
ADCY4 Polyclonal specifically detects ADCY4 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
ADCY4 | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
Q8NFM4 | |
ADCY4 | |
This antibody was developed against a recombinant protein corresponding to amino acids: FAHLSHGDSPVSTSTPLPEKTLASFSTQWSLDRSRTPRGLDDELDTGDAKFFQVIEQLNSQKQWKQSKDFNPLTLYFREKEMEKEYRLSAIP | |
25 μL | |
Lipid and Metabolism | |
196883 | |
Human | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
AC4, adenylate cyclase 4, adenylate cyclase type 4, Adenylate cyclase type IV, Adenylyl cyclase 4, ATP pyrophosphate-lyase 4, EC 4.6.1, EC 4.6.1.1 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction