Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ADCY4 Rabbit anti-Human, Mouse, Rat, Polyclonal, FabGennix

Catalog No. 502208101
Encompass
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-8101 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-8101 Supplier Fabgennix Inc Supplier No. PAC401AP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ADCY4 Polyclonal antibody specifically detects ADCY4 in Human, Mouse, Rat samples. It is validated for ELISA, Immunocytochemistry, Immunohistochemistry, Immunomicroscopy, Immunoprecipitation, Western Blot

Specifications

Antigen ADCY4
Applications ELISA, Immunocytochemistry, Immunohistochemistry, Immunomicroscopy, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.5-1.5 mg/mL
Conjugate Unconjugated
Formulation proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8
Gene Adcy4
Gene Accession No. P26770, Q8NFM4, Q91WF3
Gene Alias A cyclase IV; AC4; Adcy4; adenylate cyclase 4; adenylate cyclase type 4; Adenylate cyclase type IV; adenylyl cyclase 4; ATP pyrophosphate-lyase 4; KIAA4004; mKIAA4004
Host Species Rabbit
Immunogen Synthetic peptide corresponding to unique amino acid sequence on rat Particulate Adenylyl Cyclase 4 protein.
Purification Method Affinity chromatography
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 104110, 196883, 54223
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.