Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ADCY7 Rabbit anti-Bovine, Human, Mouse, Rat, Polyclonal, FabGennix

Catalog No. 502208110
Encompass
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-8110 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-8110 Supplier Fabgennix Inc Supplier No. PAC701AP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ADCY7 Polyclonal antibody specifically detects ADCY7 in Bovine, Human, Mouse, Rat samples. It is validated for ELISA, Immunocytochemistry, Immunohistochemistry, Immunomicroscopy, Immunoprecipitation, Western Blot

Specifications

Antigen ADCY7
Applications ELISA, Immunocytochemistry, Immunohistochemistry, Immunomicroscopy, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.5-1.5 mg/mL
Conjugate Unconjugated
Formulation proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8
Gene ADCY7
Gene Accession No. P51828, P51829, Q29450
Gene Alias AA407758; AC7; ADCY7; adenylate cyclase 7; adenylate cyclase type 7; Adenylate cyclase type VII; adenylyl cyclase 7; adenylyl cyclase type VII; ATP pyrophosphate-lyase 7; FLJ36387; KIAA0037
Host Species Rabbit
Immunogen Synthetic peptide corresponding to unique amino acid sequence on mouse Particulate Adenylyl Cyclase 7 protein.
Purification Method Affinity chromatography
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 113, 11513, 281603, 84420
Target Species Bovine, Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.