Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ADCY9 Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS3926R

Description
Adenylate cyclase is a membrane bound enzyme that catalyses the formation of cyclic AMP from ATP. It is regulated by a family of G protein-coupled receptors, protein kinases, and calcium. The type 9 adenylyl cyclase is a widely distributed adenylyl cyclase, and it is stimulated by beta-adrenergic receptor activation but is insensitive to forskolin, calcium, and somatostatin.Specifications
ADCY9 | |
Polyclonal | |
Unconjugated | |
ADCY9 | |
AC9; ACIX; ACTP10; Adcy9; adenylate cyclase 9; Adenylate cyclase type 9; Adenylate cyclase type IX; Adenylyl cyclase 9; Adenylyl cyclase type 10; ATP pyrophosphate-lyase 9; AW125421; D16Wsu65e; KIAA0520; mKIAA0520; type IX ATP pyrophosphate-lyase | |
Rabbit | |
Protein A | |
RUO | |
115 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
O60503 | |
ADCY9 | |
KLH conjugated synthetic peptide derived from human ADCY9. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction