Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ADD1/ADD2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA536044
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA536044 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA536044 Supplier Invitrogen™ Supplier No. PA536044
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody detects endogenous protein at a molecular weight of 81 kDa. Purity is >95% by SDS-PAGE.

Membrane-cytoskeleton-associated protein that promotes the assembly of the spectrin-actin network. Binds to calmodulin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ADD1/ADD2
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene ADD2
Gene Accession No. P35611, P35612, Q05764, Q63028, Q9QYB8, Q9QYC0
Gene Alias 2900072M03Rik; ADD1; Add2; Add97; ADDA; ADDB; adducin 1; adducin 1 (alpha); adducin 2; adducin 2 (beta); adducin beta; Adducin1; Adducin-1; adducin-63; Adducin-alpha; Adducin-beta; AI256389; alpha Adducin; alpha-ADD; alpha-adducin; beta Adducin; beta-ADD; beta-adducin; erythrocyte adducin alpha subunit; erythrocyte adducin subunit alpha; Erythrocyte adducin subunit beta; snoRNA MBII-396
Gene Symbols ADD1, ADD2
Host Species Rabbit
Immunogen Synthetic peptide corresponding to amino acids 700-750 of human Adducin alpha.
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11518, 11519, 118, 119, 24170, 24171
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.