Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ADE2 Antibody (OTI1B4), PerCP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271610PCP
Description
ADE2 Monoclonal specifically detects ADE2 in Human, Mouse, Rat samples. It is validated for Flow Cytometry.Specifications
| ADE2 | |
| Monoclonal | |
| PerCP | |
| PBS with 0.05% Sodium Azide | |
| PAICS | |
| Full length recombinant protein of human ADE2 (NP_006443) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG2b |
| Flow Cytometry | |
| OTI1B4 | |
| Flow Cytometry | |
| ADE2, AIR carboxylase, AIRCADE2H1, DKFZp781N1372, MGC1343, MGC5024, multifunctional protein ADE2, PAIS, phosphoribosylaminoimidazole carboxylase, phosphoribosylaminoimidazolesuccinocarboxamide synthetase, SAICAR synthetase | |
| Mouse | |
| Affinity Purified | |
| Stem Cell Markers | |
| 10606.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction