Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific Chemicals Adefovir dipivoxil
Specifications
Format | Powder |
---|---|
Molecular Weight (g/mol) | 501.47 |
Product Type | Adefovir dipivoxil |
Source | synthetic source |
Content And Storage | Freezer -20°C |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
AC463080010
|
Thermo Scientific Chemicals
463080010 |
1 g | N/A | N/A | N/A | ||||
AC463080100
|
Thermo Scientific Chemicals
463080100 |
10 g | N/A | N/A | N/A | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Powder | |
Adefovir dipivoxil | |
Freezer -20°C | |
normal conditions |
501.47 | |
synthetic source | |
98.0 to 102.0 % (Titration with HClO4) | |
White to almost white |
Safety and Handling
ShelfLife : 5 years
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title