Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Adenine Nucleotide Translocase 1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP159594
Description
Adenine Nucleotide Translocase 1 Polyclonal specifically detects Adenine Nucleotide Translocase 1 in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Adenine Nucleotide Translocase 1 | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
AAC1, Adenine nucleotide translocator 1, ADP, ADP/ATP translocase 1, ANT 1, ANT1adenine nucleotide translocator 1 (skeletal muscle), ATP carrier protein 1, ATP carrier protein, heart/skeletal muscle, ATP carrier protein, heart/skeletal muscle isoform T1, heart/skeletal muscle ATP/ADP translocator, PEO2, PEO3, solute carrier family 25 (mitochondrial carrier; adenine nucleotidetranslocator), member 4, Solute carrier family 25 member 4, T1ANT | |
Rabbit | |
Affinity purified | |
RUO | |
291 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
Q05962 | |
SLC25A4 | |
Synthetic peptides corresponding to SLC25A4 (solute carrier family 25 (mitochondrial carrier; adenine nucleotide translocator), member 4) The peptide sequence was selected from the N terminal of SLC25A4. Peptide sequence LLQVQHASKQISAEKQYKGIIDCVVRIPKEQGFLSFWRGNLANVIRYFPT The peptide sequence for this immunogen was taken from within the described region. | |
100 μL | |
Primary | |
Expected identity based on immunogen sequence: Bovine: 100%; Canine: 100%; Goat: 100%; Human: 100%; Mouse: 100%; Rat: 100%; Sheep: 100%; Chicken: 92%; Xenopus: 84%; Zebrafish: 78%. | |
Human, Rat, Bovine, Canine, Equine, Equine, Guinea Pig, Goat, Rabbit, Sheep, Zebrafish | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction