Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Adenine Nucleotide Translocase 1 Rabbit anti-Mouse, Polyclonal, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$501.50

Specifications

Antigen Adenine Nucleotide Translocase 1
Dilution Western Blot 1.0 ug/ml
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB120095
SDP
View Documents
Novus Biologicals
NBP310974100UL
100 μg
Each of 1 for $501.50
Only null left
Add to Cart
 
Description

Description

Adenine Nucleotide Translocase 1 Polyclonal specifically detects Adenine Nucleotide Translocase 1 in Mouse samples. It is validated for Western Blot.
Specifications

Specifications

Adenine Nucleotide Translocase 1
Western Blot
Unconjugated
Rabbit
Mouse
AAC1, Adenine nucleotide translocator 1, ADP, ADP/ATP translocase 1, ANT 1, ANT1adenine nucleotide translocator 1 (skeletal muscle), ATP carrier protein 1, ATP carrier protein, heart/skeletal muscle, ATP carrier protein, heart/skeletal muscle isoform T1, heart/skeletal muscle ATP/ADP translocator, PEO2, PEO3, solute carrier family 25 (mitochondrial carrier; adenine nucleotidetranslocator), member 4, Solute carrier family 25 member 4, T1ANT
The immunogen is a synthetic peptide directed towards the N terminal region of mouse Adenine Nucleotide Translocase 1 (NP_031476.3). Peptide sequence AVSKTAVAPIERVKLLLQVQHASKQISAEKQYKGIIDCVVRIPKEQGFLS
Affinity purified
Western Blot 1.0 ug/ml
Polyclonal
Purified
RUO
PBS buffer, 2% sucrose
291
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.