Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Adeno-associated virus 9 Mouse anti-Adenovirus, Unconjugated, Clone: HL2370, MilliporeSigma™

Catalog No. MABF23252MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF23252MI 25 μL
MABF23251MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABF23252MI Supplier MilliporeSigma Supplier No. MABF232525UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Adeno-associated virus 9 Monoclonal antibody specifically detects Adeno-associated virus 9 in Adenovirus samples. It is validated for Dot Blot, ELISA, Neutralizing.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Adeno-associated virus 9
Applications Dot Blot, ELISA, Neutralization
Classification Monoclonal
Clone HL2370
Conjugate Unconjugated
Gene Alias Adeno-associated virus serotype 9;AAV9
Host Species Mouse
Immunogen Recombinant Adeno-associated virus serotype 9 capsid protein.
Purification Method Protein G Purified
Quantity 25 μL
Research Discipline Inflammation and Immunology
Primary or Secondary Primary
Test Specificity Clone HL2370 is a mouse monoclonal antibody that specifically detects AAV9 capsid protein.
Target Species Adenovirus
Content And Storage Stable for 1 year at -20°C from date of receipt.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.