Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Adeno-associated Virus 9 Mouse anti-Virus, Unconjugated, Clone: HL2372, MilliporeSigma™

Catalog No. MABF23091MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF23091MI 100 μL
MABF23092MI 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABF23091MI Supplier MilliporeSigma Supplier No. MABF2309100UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Adeno-associated Virus 9 Monoclonal antibody specifically detects Adeno-associated Virus 9 in Virus samples. It is validated for Dot Blot, ELISA, Neutralizing.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Adeno-associated Virus 9
Applications Dot Blot, ELISA, Neutralization
Classification Monoclonal
Clone HL2372
Conjugate Unconjugated
Gene Alias AAV9;Adeno-associated viruses serotype 9
Host Species Mouse
Immunogen Adeno-associated virus 9 capsids.
Purification Method Protein G Purified
Quantity 100 μL
Research Discipline Inflammation and Immunology
Primary or Secondary Primary
Test Specificity Clone HL2372 is a mouse monoclonal antibody that detects adenoassociated viruses (AAV9 and AAV8). It binds only to intact capsids and not to linear viral proteins.
Target Species Virus
Content And Storage Stable for 1 year at -20°C from date of receipt.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.