Learn More
Adeno-associated Virus 9 Mouse anti-Virus, Unconjugated, Clone: HL2372, MilliporeSigma™
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Antigen | Adeno-associated Virus 9 |
| Applications | Dot Blot, ELISA, Neutralization |
| Classification | Monoclonal |
| Clone | HL2372 |
| Conjugate | Unconjugated |
| Gene Alias | AAV9;Adeno-associated viruses serotype 9 |
| Host Species | Mouse |
| Immunogen | Adeno-associated virus 9 capsids. |
| Purification Method | Protein G Purified |
| Quantity | 25 μL |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.