Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Adenomatous Polyposis Coli Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501985577
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-5577 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-5577 Supplier Bioss Supplier No. BS3680R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene encodes a tumor suppressor protein that acts as an antagonist of the Wnt signaling pathway. It is also involved in other processes including cell migration and adhesion, transcriptional activation, and apoptosis. Defects in this gene cause familial adenomatous polyposis (FAP), an autosomal dominant pre-malignant disease that usually progresses to malignancy. Disease-associated mutations tend to be clustered in a small region designated the mutation cluster region (MCR) and result in a truncated protein product.

Specifications

Antigen Adenomatous Polyposis Coli
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Apc
Gene Accession No. P25054
Gene Alias adenomatosis polyposis coli; adenomatosis polyposis coli tumor suppressor; adenomatous polyposis coli; Adenomatous polyposis coli protein; AI047805; APC; APC, WNT signaling pathway regulator; AU020952; AW124434; BTPS2; CC1; Deleted in polyposis 2.5; DP2; DP2.5; DP3; FAP; FPC; GS; mAPC; Min; multiple intestinal neoplasia; PPP1R46; Protein APC; protein phosphatase 1, regulatory subunit 46; RATAPC; truncated adenomatosis polyposis coli; WNT signaling pathway regulator
Gene Symbols Apc
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human APC.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 324
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.