Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Adenosine 3',5'-cyclic monophosphate, 99+%

Catalog No. AC225800010
Change view
Click to view available options
Quantity:
500 mg
1 g
5 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC225800010 1 g Glass bottle
AC225805000 500 mg Glass bottle
AC225800050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC225800010 Supplier Thermo Scientific Chemicals Supplier No. 225800010
Add to Cart

CAS: 60-92-4 | C10H12N5O6P | 329.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60-92-4
Molecular Formula C10H12N5O6P
Molecular Weight (g/mol) 329.21
MDL Number MFCD00005845
InChI Key IVOMOUWHDPKRLL-YPLCUDRINA-N
Synonym camp, cyclic amp, adenosine 3',5'-cyclic monophosphate, 3',5'-cyclic amp, adenosine 3',5'-phosphate, adenosine 3',5'-cyclophosphate, cyclic 3',5'-amp, adenosine cyclic monophosphate, adenosine-3',5'-cyclophosphate, cyclic adenosine monophosphate
PubChem CID 6076
ChEBI CHEBI:17489
SMILES NC1=C2N=CN([C@@H]3O[C@@H]4COP(O)(=O)O[C@H]4[C@H]3O)C2=NC=N1

Specifications

CAS Min % 99.0
Absorbance lambda max. 260nm,pH 7,0.77 to 0.83 (A250/A260),0.13 to 0.19 (A280/A260)
CAS Max % 100.0
Melting Point 260.0°C
Assay Percent Range 99% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Beilstein 26, IV, 3618
Merck Index 15, 2701
Specific Rotation − 53.70
Solubility Information Solubility in water: 50mg/mL
IUPAC Name (4aR,6R,7R,7aS)-6-(6-amino-9H-purin-9-yl)-2,7-dihydroxy-hexahydro-2λ⁵-furo[3,2-d][1,3,2]dioxaphosphinin-2-one
Formula Weight 329.2
Percent Purity 99+%
Chemical Name or Material Adenosine 3′,5′-cyclic monophosphate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 200-492-9

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.