Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Adenosine-5'-Diphosphate Trisodium Salt, MP Biomedicals

Catalog No. ICN10004905 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10004905 5 g
ICN10004901 1 g
ICN10004910 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN10004905 Supplier MP Biomedicals, Inc Supplier No. 0210004905
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2092-65-1
Molecular Formula C10H15N5Na3O10P2
Molecular Weight (g/mol) 496.172
InChI Key VJZLETZZUNQROW-MSQVLRTGSA-N
Synonym adenosine-5'-diphosphate trisodium salt
PubChem CID 131875859
IUPAC Name [(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphono hydrogen phosphate;sodium
SMILES C1=NC2=C(C(=N1)N)N=CN2C3C(C(C(O3)COP(=O)(O)OP(=O)(O)O)O)O.[Na].[Na].[Na]

Specifications

Quantity 5 g
Formula Weight 493.2
Chemical Name or Material Adenosine-5'-Diphosphate Trisodium Salt
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.