Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Adenosine-5'-Triphosphate Disodium Salt Hydrate MP Biomedicals

Catalog No. ICN15026605 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
25 g
100 g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15026605 5 g
ICN15026601 1 g
MP219461410 10 g
ICN19461425 25 g
ICN15026680 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. ICN15026605 Supplier MP Biomedicals, Inc Supplier No. 0215026605
Add to Cart
Add to Cart

Adenosine-5'-triphosphate Disodium Salt Hydrate Cell culture reagent, ≥96%, White Powder

  • Adenosine 5'-triphosphate (ATP) and its phosphate bonds are the basic components of energy exchange in many biological systems.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51963-61-2
Molecular Formula C10H20N5Na2O16P3
Molecular Weight (g/mol) 605.19
MDL Number MFCD00150754
InChI Key MWEQTWJABOLLOS-MVHBFMHHNA-L
PubChem CID 134129625
SMILES O.O.O.[Na+].[Na+].NC1=C2N=CN([C@@H]3O[C@H](COP(O)(=O)OP([O-])(=O)OP(O)([O-])=O)[C@@H](O)[C@H]3O)C2=NC=N1

Specifications

Color White
Physical Form Powder
Quantity 5 g
Synonym adenosine 5'-triphosphate disodium salt trihydrate, adenosine 5'-triphosphate disodium salt, adenosine 5'-triphosphate disodium salt trihydrate, disodium salt
IUPAC Name disodium;[[[(2R,3S,4S,5R)-5-(6-aminopurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy-hydroxyphosphoryl]oxy-oxidophosphoryl] hydrogen phosphate;trihydrate
Formula Weight 551.2 (Anhydrous)
Chemical Name or Material Adenosine-5'-Triphosphate Disodium Salt Hydrate, Hydrate
Recommended Storage Store at 0°C.
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.