Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Adenosine A1R Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 6 publications
Supplier: Novus Biologicals NB300549
Description
Adenosine A1R Polyclonal specifically detects Adenosine A1R in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
Adenosine A1R | |
Polyclonal | |
Unconjugated | |
PBS and 1 mg/ml BSA with 0.05% Sodium Azide | |
adenosine A1 receptor, ADORA 1, RDC7adenosine receptor A1 | |
Rabbit | |
37 kDa | |
100 μL | |
Primary | |
Detects Adenosine Receptor A1. This does not detect other AR subtypes. | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
1 mg/mL | |
Western Blot 1:500, Flow Cytometry 3 - 5 ug/ 10^6 cells, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 2 ug/ml, Immunohistochemistry-Paraffin 1:50 - 1:200, Immunohistochemistry-Frozen | |
P25099 | |
ADORA1 | |
Synthetic peptide corresponding to residues C(309) Q P K P P I D E D L P E E K A E D(326) of rat Adenosine Receptor A1. | |
Affinity Purified | |
RUO | |
134 | |
Human, Mouse, Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction