Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Adenosine A1R Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$382.00 - $610.00

Specifications

Antigen Adenosine A1R
Applications Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP258443
SDP
View Documents
Novus Biologicals
NBP258443
100 μL
Each for $610.00
Only null left
Add to Cart
 
NB403205
SDP
View Documents
Novus Biologicals
NBP25844325UL
25 μL
Each for $382.00
Only null left
Add to Cart
 
Description

Description

Adenosine A1R Polyclonal specifically detects Adenosine A1R in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

Adenosine A1R
Polyclonal
Rabbit
Apoptosis, GPCR, Growth and Development, Immunology, Innate Immunity, Neuronal Cell Markers, Signal Transduction, Vision
adenosine A1 receptor, ADORA 1, RDC7adenosine receptor A1
ADORA1
IgG
Affinity Purified
Immunocytochemistry, Immunofluorescence
Unconjugated
RUO
Human
134
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:NNLSAVERAWAANGSMGEPVIKCEFEKVISMEYMVYFN
Primary
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.