Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Adenosine A2aR/A2bR Antibody (826037), Alexa Fluor™ 532, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB94973X
Description
Adenosine A2aR/A2bR Monoclonal antibody specifically detects Adenosine A2aR/A2bR in Human samples. It is validated for Flow CytometrySpecifications
Adenosine A2aR/A2bR | |
Monoclonal | |
Alexa Fluor 532 | |
50mM Sodium Borate | |
NS0 mouse myeloma cell line transfected with human Adenosine A2aR, Met1-Ser412, Accession # P29274 | |
0.1 mL | |
Primary | |
Store at 4C in the dark. | |
IgG2b |
Flow Cytometry | |
826037 | |
Flow Cytometry | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction