Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Adenosine A2aR Antibody (7F6-G5-A2), Alexa Fluor™ 532, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP139474AF532
Description
Adenosine A2aR Monoclonal specifically detects Adenosine A2aR in Human, Mouse, Rat, Porcine, Canine, Guinea Pig, Rabbit, Sheep samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Flow (Intracellular).Specifications
Adenosine A2aR | |
Monoclonal | |
Alexa Fluor 532 | |
50 mM sodium borate with 0.05% sodium azide | |
ADORA2A | |
Full length recombinant human Adenosine A2a R. [UniProtNo. P29274] | |
Primary | |
Human, Mouse, Rat, Pig, Canine, Guinea Pig, Rabbit, Sheep | |
IgG2a |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
7F6-G5-A2 | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Flow (Intracellular) | |
adenosine A2 receptor, adenosine A2a receptor, adenosine receptor A2a, adenosine receptor subtype A2a, ADORA 2, ADORA2, hA2aR, RDC8 | |
Mouse | |
0.1 mL | |
135 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction