Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Adenosine A2aR Antibody (7F6-G5-A2), mFluor Violet 450 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP139474MFV450
Description
Adenosine A2aR Monoclonal antibody specifically detects Adenosine A2aR in Human, Mouse, Rat, Porcine, Canine, Guinea Pig, Rabbit, Sheep samples. It is validated for Flow Cytometry, Flow (Intracellular)Specifications
Adenosine A2aR | |
Monoclonal | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Pig, Canine, Guinea Pig, Rabbit | |
Purified |
Flow Cytometry | |
7F6-G5-A2 | |
Flow Cytometry, Flow (Intracellular) | |
adenosine A2 receptor, adenosine A2a receptor, adenosine receptor A2a, adenosine receptor subtype A2a, ADORA 2, ADORA2, hA2aR, RDC8 | |
Full length recombinant human Adenosine A2a R. [UniProt# P29274] | |
0.1 mL | |
Apoptosis, GPCR, Immunology, Innate Immunity | |
135 | |
Store at 4C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction