Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Adenosine A2b R Antibody (2683F), mFluor Violet 500 SE, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals FAB104442MFV500
Description
Adenosine A2b R Monoclonal antibody specifically detects Adenosine A2b R in Human samples. It is validated for Flow CytometrySpecifications
Adenosine A2b R | |
Monoclonal | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Rabbit | |
0.1 mL | |
Immunology, Signal Transduction | |
136 | |
Store at 4C in the dark. |
Flow Cytometry | |
2683F | |
Flow Cytometry | |
adenosine A2b receptor, adenosine receptor A2b, ADORA2 | |
Human Adenosine A2b R synthetic peptide, Accession # NP_000667 | |
RUO | |
Primary | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction