Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Adenosine A2b R Antibody, mFluor Violet 450 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP241312MFV450
Description
Adenosine A2b R Polyclonal antibody specifically detects Adenosine A2b R in Human, Mouse, Rat samples. It is validated for Flow CytometrySpecifications
| Adenosine A2b R | |
| Polyclonal | |
| Flow Cytometry | |
| adenosine A2b receptor, adenosine receptor A2b, ADORA2 | |
| Antibody was raised against a 19 amino acid peptide near the carboxy terminus of human ADORA2B. The immunogen is located within the last 50 amino acids of ADORA2B. Amino Acid Squence: QADVKSGNGQAGVQPA | |
| 0.1 mL | |
| Immunology, Signal Transduction | |
| 136 | |
| Store at 4C in the dark. | |
| IgG |
| Flow Cytometry | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction