Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Adenosine Deaminase/ADA Antibody, Biotin, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP300364B
Description
Adenosine Deaminase/ADA Polyclonal specifically detects Adenosine Deaminase/ADA in Human samples. It is validated for Western Blot, ELISA.Specifications
Adenosine Deaminase/ADA | |
Polyclonal | |
Western Blot, ELISA | |
ADA1, adenine deaminase, Adenosine aminohydrolase, adenosine deaminase, EC 3.5.4.4 | |
Produced in rabbits immunized with purified, recombinant Adenosine Deaminase/ADA (P00813; Met1-Leu363). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot, ELISA | |
Biotin | |
PBS | |
Rabbit | |
Affinity purified | |
RUO | |
100 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction