Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Adenosine Deaminase/ADA Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$203.51 - $530.02

Specifications

Antigen Adenosine Deaminase/ADA
Applications Western Blot, Immunofluorescence, KnockDown
Classification Polyclonal
Conjugate Unconjugated
Form Purified
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB093038
SDP
View Documents
Novus Biologicals
NBP30346920UL
20 μL
Each for $203.51
Only null left
Add to Cart
 
NB093037
SDP
View Documents
Novus Biologicals
NBP303469100UL
100 μL
Each for $530.02
Only null left
Add to Cart
 
Description

Description

Adenosine Deaminase/ADA Polyclonal specifically detects Adenosine Deaminase/ADA in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Knockout Validated.
Specifications

Specifications

Adenosine Deaminase/ADA
Polyclonal
Purified
RUO
PBS with 50% glycerol, pH7.3.
100
IgG
Affinity purified
Western Blot, Immunofluorescence, KnockDown
Unconjugated
Rabbit
Human, Mouse, Rat
ADA1, adenine deaminase, Adenosine aminohydrolase, adenosine deaminase, EC 3.5.4.4
Recombinant fusion protein containing a sequence corresponding to amino acids 1-363 of human Adenosine Deaminase/ADA (NP_000013.2).
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.