Learn More
Invitrogen™ Adenosine Deaminase Polyclonal Antibody

Description
This antibody has specificity for Human Adenosine Deaminase/ADA.
Specifications
Specifications
| Antigen | Adenosine Deaminase |
| Applications | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.03% ProClin 300; pH 7 |
| Gene | ADA |
| Gene Accession No. | P00813 |
| Gene Alias | ada; ADA1; adenosine aminohydrolase; Adenosine deaminase; RP11-61L14.5; zgc:92028 |
| Gene Symbols | ADA |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.