Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Adenosine Deaminase Recombinant Rabbit Monoclonal Antibody (5R10B4)
GREENER_CHOICE

Catalog No. PIMA538137
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA538137 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA538137 Supplier Invitrogen™ Supplier No. MA538137
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Positive Samples: Jurkat, Mouse stomach Immunogen sequence: NRLRQENMHF EICPWSSYLT GAWKPDTEHA VIRLKNDQAN YSLNTDDPLI FKSTLDTDYQ MTKRDMGFTE EEFKRLNINA AKSSFLPEDE KRELLDLLYK A.

Catalyzes the hydrolytic deamination of adenosine and 2-deoxyadenosine. Plays an important role in purine metabolism and in adenosine homeostasis. Modulates signaling by extracellular adenosine, and so contributes indirectly to cellular signaling events. May act as a positive regulator of T-cell coactivation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Adenosine Deaminase
Applications ELISA, Western Blot
Classification Recombinant Monoclonal
Clone 5R10B4
Concentration 3 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ADA
Gene Accession No. P00813, P03958
Gene Alias ada; ADA1; adenosine aminohydrolase; Adenosine deaminase; RP11-61L14.5; zgc:92028
Gene Symbols ADA
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 250-350 of human Adenosine Deaminase (ADA) (NP_0000132).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100, 11486
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.