Learn More
Invitrogen™ Adenosine Kinase Polyclonal Antibody

Description
Recommended positive controls: A431, HepG2, HeLa, mouse liver, rat liver. Predicted reactivity: Mouse (89%), Rat (87%), Chicken (83%), Rhesus Monkey (99%), Bovine (93%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | Adenosine Kinase |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.28 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 20% glycerol and 0.025% ProClin 300; pH 7 |
| Gene | ADK |
| Gene Accession No. | P55263, P55264, Q64640 |
| Gene Alias | 2310026J05Rik; 5033405D03Rik; Adenosine 5'-phosphotransferase; adenosine kinase; ADK; AI255373; AI987814; AK; testicular tissue protein Li 14 |
| Gene Symbols | ADK |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.