Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Adenosine Kinase Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA583416
Description
Immunogen sequence: LFGMGNPLLD ISAVVDKDFL DKYSLKPNDQ ILAEDKHKEL FDELVKKFKV EYHAGGSTQN SIKVAQWMIQ QPHK.
The protein encoded by this gene catalyzes the addition of glucose monomers to the growing glycogen molecule through the formation of alpha-1,4-glycoside linkages. Mutations in this gene are associated with muscle glycogen storage disease. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
Specifications
Adenosine Kinase | |
Polyclonal | |
Unconjugated | |
ADK | |
2310026J05Rik; 5033405D03Rik; Adenosine 5'-phosphotransferase; adenosine kinase; ADK; AI255373; AI987814; AK; testicular tissue protein Li 14 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
11534, 132, 25368 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P55263, P55264, Q64640 | |
ADK | |
Recombinant protein corresponding to Human ADK. Recombinant protein control fragment (Product #RP-97139). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction