Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Adenosine, MP Biomedicals™

Catalog No. MP219980305 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 g
25 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP219980305 5 g
MP219980325 25 g
MP219980391 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. MP219980305 Supplier MP Biomedicals, Inc Supplier No. 0219980305
Add to Cart
Add to Cart

Comprised of a molecule of adenine attached to a ribose sugar molecule moiety via a β-N9-glycosidic bond. Adenosine, MP Biomedicals is a purine nucleoside that is an endogenous neurotransmitter at adenosine receptors.

  • Cardioprotective effects may relate to activation of A1 adenosine receptors and it is used for the treatment or prevention of cardiac arrhythmias (irregular heartbeat)
  • The antiplatelet and anti-inflammatory actions of adenosine appear to be mediated via the A2 adenosine receptor
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58-61-7
Molecular Formula C10H13N5O4
Molecular Weight (g/mol) 267.25
MDL Number MFCD00005752
InChI Key OIRDTQYFTABQOQ-KQYNXXCUSA-N
Synonym adenosine, adenocard, adenoscan, adenine riboside, adenosin, beta-d-adenosine, nucleocardyl, boniton, sandesin, myocol
PubChem CID 60961
ChEBI CHEBI:16335
IUPAC Name (2R,3R,4S,5R)-2-(6-amino-9H-purin-9-yl)-5-(hydroxymethyl)oxolane-3,4-diol
SMILES NC1=C2N=CN([C@@H]3O[C@H](CO)[C@@H](O)[C@H]3O)C2=NC=N1

Specifications

Melting Point 233°C to 238°C
Quantity 5 g
Optical Rotation −68.0° to −72.0° (c=2, NaOH 1:20)
Formula Weight 267.2
Percent Purity 99.0 to 101.0%
Grade USP
Physical Form Powder
Recommended Storage −20°C
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.