Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Adenosylhomocysteinase/AHCY Antibody (13H8), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | Adenosylhomocysteinase/AHCY |
---|---|
Clone | 13H8 |
Dilution | Western Blot 1:500-1:2000, ELISA |
Applications | Western Blot, ELISA |
Classification | Monoclonal |
Description
Adenosylhomocysteinase/AHCY Monoclonal antibody specifically detects Adenosylhomocysteinase/AHCY in Human samples. It is validated for Western Blot, ELISASpecifications
Adenosylhomocysteinase/AHCY | |
Western Blot 1:500-1:2000, ELISA | |
Monoclonal | |
Purified | |
RUO | |
Human | |
adenosylhomocysteinase, AdoHcyase, EC 3.3.1.1, S-adenosylhomocysteine hydrolase, S-adenosyl-L-homocysteine hydrolase, SAHHadoHcyase | |
A synthesized peptide derived from Human Adenosylhomocysteinase/AHCY [UniProt P23526] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
13H8 | |
Western Blot, ELISA | |
Unconjugated | |
Rabbit | |
Amino Acids Drugs and other small molecules, Endocrinology, Signal Transduction | |
PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
191 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title