Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Adenosylhomocysteinase/AHCY Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00
Specifications
Antigen | Adenosylhomocysteinase/AHCY |
---|---|
Dilution | Western Blot 0.04-0.4 μg/mL, Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:20 - 1:50 |
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
Adenosylhomocysteinase/AHCY Polyclonal antibody specifically detects Adenosylhomocysteinase/AHCY in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
Adenosylhomocysteinase/AHCY | |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Amino Acids Drugs and other smAll species molecules, Endocrinology, Signal Transduction | |
PBS (pH 7.2), 40% Glycerol | |
191 | |
IgG | |
Immunogen affinity purified |
Western Blot 0.04-0.4 μg/mL, Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
Polyclonal | |
Purified | |
RUO | |
Human | |
adenosylhomocysteinase, AdoHcyase, EC 3.3.1.1, S-adenosylhomocysteine hydrolase, S-adenosyl-L-homocysteine hydrolase, SAHHadoHcyase | |
This antibody was developed against a recombinant protein corresponding to amino acids: MEGYEVTTMDEACQEGNIFVTTTGCIDIILGRHFEQMKDDAIVCNIGHFDVEIDVKWLNENAVEKVNIKPQVDRYRLKNGRRIILLA | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title