Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Adenosylhomocysteinase/AHCY Rabbit anti-Human, Mouse, Rat, Clone: 4E9H10, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $458.50
Specifications
Antigen | Adenosylhomocysteinase/AHCY |
---|---|
Clone | 4E9H10 |
Applications | Western Blot, Immunoprecipitation |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
Adenosylhomocysteinase/AHCY Monoclonal antibody specifically detects Adenosylhomocysteinase/AHCY in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
Adenosylhomocysteinase/AHCY | |
Western Blot, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Amino Acids Drugs and other small molecules, Endocrinology, Signal Transduction | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
191 | |
IgG | |
Affinity purified |
4E9H10 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
adenosylhomocysteinase, AdoHcyase, EC 3.3.1.1, S-adenosylhomocysteine hydrolase, S-adenosyl-L-homocysteine hydrolase, SAHHadoHcyase | |
A synthetic peptide corresponding to a sequence within amino acids 300-400 of human Adenosylhomocysteinase/AHCY (P23526). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title