Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Adenovirus 5 E1A binding protein Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501987936
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-7936 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-7936 Supplier Bioss Supplier No. BS7697R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The protein encoded by this gene was first identified by its ability to bind the adenovirus E1A protein. The protein localizes to the nucleus. It functions as a transcriptional repressor, and expression of E1A inhibits this repression. Alternatively spliced transcript variants encoding different isoforms have been identified.

Specifications

Antigen Adenovirus 5 E1A binding protein
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene ZMYND11
Gene Accession No. Q15326
Gene Alias 2210402G22Rik; Adenovirus 5 E1A-binding protein; Bone morphogenetic protein receptor-associated molecule 1; BOS_13032; BRAM1; BS69; MGC111056; MRD30; Protein BS69; RP11-486H9.1; Zinc finger MYND domain-containing protein 11; zinc finger MYND-type containing 11; zinc finger, MYND domain containing 11; zinc finger, MYND-type containing 11; ZMYND11
Gene Symbols ZMYND11
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human Adenovirus 5 E1A binding protein.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10771
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.