Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Adenovirus Antibody (8C4), Janelia Fluor™ 646, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB600413JF6460.2ML
Description
Adenovirus Monoclonal specifically detects Adenovirus in Adenovirus, Virus samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Sandwich ELISA.Specifications
Adenovirus | |
Monoclonal | |
Janelia Fluor 646 | |
50 mM Sodium Borate | |
Mouse | |
0.2 mL | |
Virus | |
Purified |
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
8C4 | |
Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
Adenovirus | |
Hexon antigen of human adenovirus. | |
Adenovirus hexon specific. The immunogen is directed against a genus-specific hexon antigen, which is present in all human serotypes. This detects a different epitope than the monoclonal, NB200-425. | |
Store at 4°C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction