Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Adenovirus Antibody (M58 + M73), Novus Biologicals™

Mouse Monoclonal Antibody
$243.50 - $709.50
Specifications
Antigen | Adenovirus |
---|---|
Clone | M58 + M73 |
Applications | Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
Adenovirus Monoclonal specifically detects Adenovirus in Adenovirus, Virus samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
Adenovirus | |
Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
Unconjugated | |
Mouse | |
Virology Bacteria and Parasites | |
Adenovirus | |
IgG2a κ | |
Protein A or G purified | |
This antibody reacts with adenovirus. |
M58 + M73 | |
Monoclonal | |
Purified | |
RUO | |
Virus, Adenovirus | |
BALB/C mice were injected with adenovirus. | |
Primary | |
Store at 4°C. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title