Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Adenovirus Hexon Antibody (3G0), Alexa Fluor™ 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10065051AF488
Description
Adenovirus Hexon Monoclonal specifically detects Adenovirus Hexon in Adenovirus, Virus samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.Specifications
Adenovirus Hexon | |
Monoclonal | |
Alexa Fluor 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
Mouse | |
Protein A purified | |
RUO | |
Recognizes the hexon group antigen present in most adenovirus serotypes. NB100-65051 has known reactivity with at least 17 serotypes of Adenovirus including types 40 & 41. | |
Adenovirus, Virus | |
Purified |
ELISA, Immunocytochemistry, Immunofluorescence | |
3G0 | |
ELISA, Immunocytochemistry/Immunofluorescence | |
Hexon protein; Late protein 2; PII | |
ATCC VR5 virus | |
0.1 mL | |
Primary | |
Protein A purified | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction