Learn More
Adenovirus Hexon Antibody (3G0), Janelia Fluor™ 646, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NB10065051JF646
Catalog No. NB065051646
Only null left
Add to Cart
Description
Description
Adenovirus Hexon Monoclonal specifically detects Adenovirus Hexon in Adenovirus, Virus samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
AdenovirusHexon
Monoclonal
Janelia Fluor 646
50mM Sodium Borate with 0.05% Sodium Azide
Mouse
Protein A purified
Primary
Adenovirus, Virus
Purified
ELISA, Immunocytochemistry, Immunofluorescence
3G0
ELISA, Immunocytochemistry/Immunofluorescence
Hexon protein; Late protein 2; PII
ATCC VR5 virus
0.1 mL
Recognizes the hexon group antigen present in most adenovirus serotypes. NB100-65051 has known reactivity with at least 17 serotypes of Adenovirus including types 40 & 41.
Store at 4°C in the dark.
IgG1
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.