Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Adenovirus Hexon Antibody (7C11), Novus Biologicals™
SDP

Catalog No. p-7200080 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB017437 0.025 mg
NB10062168 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB017437 Supplier Novus Biologicals Supplier No. NB100621680.025MG
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Adenovirus Hexon Monoclonal specifically detects Adenovirus Hexon in Adenovirus samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Frozen, Immunodiffusion.

Specifications

Antigen Adenovirus Hexon
Applications ELISA, Immunodiffusion, Immunohistochemistry (Frozen), Immunohistochemistry
Classification Monoclonal
Clone 7C11
Conjugate Unconjugated
Dilution ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, Immunodiffusion
Formulation PBS with 0.09% Sodium Azide
Gene Alias Hexon protein; Late protein 2; PII
Host Species Mouse
Immunogen Hexon antigen of human and canine adenoviruses (Type 1)
Purification Method Affinity Purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity NB100-62168 reacts with human, canine, bovine, monkey and rat adenoviruses. It is very likely that it will react with all mammalian Adenoviruses.
Target Species Adenovirus
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG2a, IgM
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.