Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Adenovirus Hexon Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. p-7200224 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
0.5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB018211 0.1 mL
NB6001386 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB018211 Supplier Novus Biologicals Supplier No. NB60013860.1ML
Only null left
Add to Cart
Add to Cart

Goat Polyclonal Antibody

Adenovirus Hexon Protein Polyclonal specifically detects Adenovirus Hexon Protein in Adenovirus, Virus samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen.

Specifications

Antigen Adenovirus Hexon Protein
Applications Immunohistochemistry (Frozen), ELISA, Immunocytochemistry, Western Blot, Immunofluorescence
Classification Polyclonal
Concentration 4.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:10-1:2000, Immunohistochemistry-Frozen
Formulation Phosphate buffered saline with 0.1% Sodium Azide
Host Species Goat
Immunogen Gradient purified, disrupted native adenovirus type 5 virions.
Purification Method IgG purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Virology Bacteria and Parasites
Primary or Secondary Primary
Test Specificity Recognizes numerous Adeno proteins including hexon protein, and reacts with Adenovirus types 1, 2, 3, 5, 6, 7a, 8, 31, 40 & 41. Broad reactivity with other Adenovirus types is expected. Goat anti Adenovirus antibody does not cross-react with Parainfluenza types 1-3, Influenza A & B, RSV, hMPV and Rhinovirus type 16. Goat anti Adenovirus antibody does not react with uninfected Hep-2, LLCMK2 and Vero cells. Adenoviruses are 80-110 nm diameter, non-enveloped icosahedral viruses that can cause respiratory illness, infantile gastroenteritis and other mild disorders such as conjunctivitis. Features of Adenovirus have made it a useful tool for gene transfection and expression in mammalian cells.
Target Species Virus, Adenovirus
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.