Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Adenovirus Hexon Monoclonal Antibody (D36J)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA17328
Description
The MA1-7328 antibody reacts with Adenovirus hexon in viral and infected tissue samples. MA1-7328 has been successfully used in ELISA and Immunofluorescence applications. The MA1-7328 antibody was raised against Adenovirus hexon protein.
Hexon protein is a major coat protein of adenoviruses. Adenoviruses capsids have three principal protein components: the hexon, the penton, and the fiber. Hexon consists of three subunits together forming two major components of different morphological symmetry. A triangular top with three towers of density is superimposed on a more bulky pseudo hexagonal base.
Specifications
Adenovirus Hexon | |
Monoclonal | |
0.1 mg/mL | |
PBS with 0.1% sodium azide; pH 7.2 | |
Mouse | |
Protein A | |
RUO | |
Virus | |
Antibody | |
IgG1 |
ELISA, Immunocytochemistry | |
D36J | |
Unconjugated | |
Adeno Virus; AdV; HADV; HAdVC_gp08 | |
Adenovirus hexon protein. | |
100 μg | |
Primary | |
4°C | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction