Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Adenovirus Hexon Monoclonal Antibody (E44D)
GREENER_CHOICE

Catalog No. PIMA17333
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA17333 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA17333 Supplier Invitrogen™ Supplier No. MA17333
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The MA1-7333 antibody reacts with Adenovirus hexon in viral and infected tissue samples. MA1-7333 has been successfully used in ELISA and Immunofluorescence applications. The MA1-7333 antibody was raised against Adenovirus hexon protein.

Hexon protein is a major coat protein of adenoviruses. Adenoviruses capsids have three principal protein components: the hexon, the penton, and the fiber. Hexon consists of three subunits together forming two major components of different morphological symmetry. A triangular top with three towers of density is superimposed on a more bulky pseudo hexagonal base.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Adenovirus Hexon
Applications ELISA, Immunocytochemistry
Classification Monoclonal
Clone E44D
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.2
Gene Alias Adeno Virus; AdV; HADV; HAdVC_gp08
Host Species Mouse
Immunogen Adenovirus hexon protein.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.