Learn More
Invitrogen™ Adenovirus Polyclonal Antibody

Description
PA1-7201 detect the adenovirus. This antibody does not cross-react with parainfluenza 1-3, Influenza. A & B, RSV, hMPV, and Rhinovirus (type 16). PA1-7201 has been successfully used in immunofluorescence and ELISA procedures. PA1-7201 immunogen is gradient purified, disrupted Adenovirus type 5 virions. Store at 4ºC for 6 months. For extended storage, store at -20ºC.
Specifications
Specifications
| Antigen | Adenovirus |
| Applications | ELISA, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 5.0 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.1% sodium azide; pH 7.2 |
| Gene Alias | Adeno Virus; AdV; HADV |
| Host Species | Goat |
| Immunogen | Gradient purified, disrupted Adenovirus type 5 virions. |
| Purification Method | IgG Fraction |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.