Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Adenovirus Type 2 Fiber Monoclonal Antibody (2A6)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA134961
Description
Recommended positive controls: Adenovirus infected cells and tissues..
Adenoviruses are DNA viruses generally widespread in nature that are frequently the cause of acute upper respiratory tract infections (i.e. common colds). 49 human adenovirus serotypes have been discovered and divided into six subgroups, A to E, based on observation. They are an important cause of respiratory illness, conjunctivitis, gastroenteritis, and have more recently been recognized as a potential cause of sexually transmitted diseases. Several types have oncogenic potential though most cause self-limiting febrile illnesses characterized by inflammation of conjunctivae and the respiratory tract. The virus can be isolated from the majority of tonsils/adenoids surgically removed, indicating latent infections. It is not known how long the virus can persist in the body, or whether it is capable of reactivation after long periods. In patients experiencing immunosuppression (e.g. AIDS) it can be reactivated, causing disease. Adenovirus vectors, particularly those based on Ad5, have shown great promise for the development of gene therapy protocols and have been used for clinical purpose.
Specifications
Adenovirus Type 2 Fiber | |
Monoclonal | |
0.2 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
Mouse | |
Protein A | |
RUO | |
Virus | |
Antibody | |
IgG2a |
Western Blot, Immunocytochemistry | |
2A6 | |
Unconjugated | |
Adeno Virus; Adeno Virus 2; AdV; AdV-2; HADV; HADV-2 | |
UV irradiated Ad2 virus. | |
500 μL | |
Primary | |
4°C, do not freeze | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction