Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Adenovirus Type 2 Monoclonal Antibody (4D2), Invitrogen™

Catalog No. PIMA511222
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA511222 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA511222 Supplier Thermo Scientific Supplier No. MA511222

Mouse Monoclonal Antibody

MA5-11222 targets Adenovirus Fiber in IF and WB applications and shows reactivity with Virus samples. The MA5-11222 immunogen is uV-irradiated Ad2 virus.

Adenovirus Type 2 Monoclonal antibody specifically detects Adenovirus Type 2 in Virus samples. It is validated for Immunocytochemistry, Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Adenovirus Type 2
Applications Immunocytochemistry, Western Blot
Classification Monoclonal
Clone 4D2
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4
Gene Alias Adeno Virus; Adeno Virus 2; AdV; AdV-2; HADV; HADV-2
Host Species Mouse
Immunogen UV-irradiated Ad2 virus
Purification Method Protein A
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.