Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Adenylate Cyclase 3 Antibody, Novus Biologicals™
SDP

Catalog No. p-5055075 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP192683SS 0.025 mL
NBP192683 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP192683SS Supplier Novus Biologicals Supplier No. NBP192683SS
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 2 publications

Adenylate Cyclase 3 Polyclonal specifically detects Adenylate Cyclase 3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Adenylate Cyclase 3
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000, Immunohistochemistry 1:10000-1:20000, Immunocytochemistry/Immunofluorescence 1:10000-1:20000
Formulation 50% PBS, 50% Glycerol with 5mM Sodium Azide
Gene Accession No. O60266
Gene Alias AC3EC 4.6.1.1, AC-III, adenylate cyclase 3, adenylate cyclase type 3, Adenylate cyclase type III, Adenylyl cyclase 3, adenylyl cyclase, type III, ATP pyrophosphate-lyase 3, EC 4.6.1, KIAA0511, olfactive type
Gene Symbols ADCY3
Host Species Rabbit
Immunogen C-terminal peptide of rat ACIII, PAAFPNGSSVTLPHQVVDNP with a Cys added to the N-terminus to allow coupling to KLH.
Molecular Weight of Antigen 180 kDa
Purification Method Affinity Purified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 109
Target Species Human, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.