Learn More
Invitrogen™ Adenylate Cyclase 3 Polyclonal Antibody
Description
Adenylate Cyclase 3 Polyclonal Antibody for Western Blot, ICC/IF, IHC
Specifications
Specifications
| Antigen | Adenylate Cyclase 3 |
| Applications | Immunohistochemistry, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 5mM sodium azide |
| Gene | Adcy3 |
| Gene Accession No. | O60266, P21932, Q8VHH7 |
| Gene Alias | AC3; AC-III; ADCY3; adenylate cyclase 3; adenylate cyclase 8 (brain); adenylate cyclase type 3; Adenylate cyclase type III; adenylate cyclase, olfactive type; Adenylyl cyclase 3; adenylyl cyclase, type III; ATP pyrophosphate-lyase 3; KIAA0511; mKIAA0511; type III adenylyl cyclase |
| Gene Symbols | Adcy3 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.